Learn More
MSC Rubbermaid Tailor Metal Frame, 41 gal, Black
Catalog No. 502838239
Encompass
This decorative refuse container frame has a 41 gallon capacity and a sleek black finish. It is constructed from powder-coated steel, providing a smooth finish and a classic design, and is intended to be used with separately sold side panels.
  • Powder-coated steel construction provides a smooth finish and classic design.
  • Frame only; side panels are sold separately.
  • Designed for 41 gallon capacity refuse containers.
  • Rectangle container shape.
  • Features a black color/finish.
  • Measures 44.63 in high, 25.20 in wide, and 25.2 in long.
Catalog No. 50-283-8239 Supplier MSC Supplier No. 60900164
This decorative refuse container frame has a 41 gallon capacity and a sleek black finish. It is constructed from powder-coated steel, providing a smooth finish and a classic design, and is intended to be used with separately sold side panels.
  • Powder-coated steel construction provides a smooth finish and classic design.
  • Frame only; side panels are sold separately.
  • Designed for 41 gallon capacity refuse containers.
  • Rectangle container shape.
  • Features a black color/finish.
  • Measures 44.63 in high, 25.20 in wide, and 25.2 in long.