Learn More
Seqmatic LLC TailorMag 12 Tube PCR Magnetic Stand (version 2)
SDP
Catalog No. NC1482181
Encompass
Optimized for low volume applications (as low as 5 μL total volume)- Strong magnets enable complete removal of sample with no dead volume- Allows processing up to 12 samples in individual PCR tubes or strips-Compatible with multichannel pipetting- Anti-flip structure and rubber feet provide extra stability- Light color background improves visibility of bead pellets
Catalog No. NC1482181 Supplier Seqmatic LLC Supplier No. TM710
Optimized for low volume applications (as low as 5 μL total volume)- Strong magnets enable complete removal of sample with no dead volume- Allows processing up to 12 samples in individual PCR tubes or strips-Compatible with multichannel pipetting- Anti-flip structure and rubber feet provide extra stability- Light color background improves visibility of bead pellets