Learn More
Andwin Scientific Tube Closure Polyethylene Thumb Cap, Yellow, 16 mm for 16 mm Glass Culture Tubes, 1000/pk
SDP
Catalog No. NC1781375 Shop All Fisherbrand Products
Encompass_Preferred
These caps are ideal for resealing glass test tubes and glass culture tubes. Economical way to recap glass tubes, regardless of manufacturer. Caps can be written on with indelible ink for easy sample identification. Prevents spills and evaporation by employing a liquid tight seal.
Catalog No. NC1781375
These caps are ideal for resealing glass test tubes and glass culture tubes. Economical way to recap glass tubes, regardless of manufacturer. Caps can be written on with indelible ink for easy sample identification. Prevents spills and evaporation by employing a liquid tight seal.