Learn More
Medchemexpress LLC HY-122190 10mg Medchemexpress, TAK-071 CAS:1820812-16-5 Purity:>98%
SDP
Catalog No. 502080982
Encompass_Preferred
Medchemexpress, HY-122190 10mg TAK-071 CAS:1820812-16-5 TAK-071 is a novel, potent and highly selective muscarinic acetylcholine receptor 1 (M1R) positive allosteric modulator. EC50 of TAK-071 M1R agonist activities is 520 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0982 Supplier Medchemexpress LLC Supplier No. HY12219010MG
Medchemexpress, HY-122190 10mg TAK-071 CAS:1820812-16-5 TAK-071 is a novel, potent and highly selective muscarinic acetylcholine receptor 1 (M1R) positive allosteric modulator. EC50 of TAK-071 M1R agonist activities is 520 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.