Learn More
TARGETMOL CHEMICALS INC TAK-137 1MG
SDP
Catalog No. 502879995
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. TAK-137 is an AMPA receptor enhancer with antidepressant effects. TAK-137 has the ability to improve cognition and could be used to study schizophrenia. Purity 99.27%
Catalog No. 50-287-9995 Supplier TARGETMOL CHEMICALS INC Supplier No. T708801MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. TAK-137 is an AMPA receptor enhancer with antidepressant effects. TAK-137 has the ability to improve cognition and could be used to study schizophrenia. Purity 99.27%