Learn More
Apexbio Technology LLC TAK-242 243984-11-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A38505.1
Encompass_Preferred
TAK-242 (CAS 243984-11-4) also known as resatorvid is a cyclohexene derivative that functions as a selective inhibitor of Toll-like receptor 4 (TLR4) signaling Mechanistically TAK-242 binds specifically to the intracellular domain of TLR4 disrupting its interaction with downstream adaptor proteins and thus suppressing activation of inflammatory signal pathways triggered by lipopolysaccharide (LPS) In vitro TAK-242 inhibits LPS-induced production of nitric oxide TNF- and interleukin-6 in macrophages (IC50 1 1 11 nM) Preclinical animal models indicate its potential usefulness in mitigating inflammatory responses associated with neuropsychiatric conditions triggered by stress
Catalog No. 50-000-64029