Learn More
Medchemexpress LLC HY-11109 100mg , TAK-242 Resatorvid CAS:243984-11-4 Purity:98%
SDP
Catalog No. 501492665
Encompass_Preferred
Medchemexpress, HY-11109 100mg TAK-242 Resatorvid CAS:243984-11-4 Formula:C15H17ClFNO4S TLR4 Purity:98% TAK-242 is a potent TLR4 signaling inhibitor, selectively inhibits the TLR4-mediated production of cytokines and NO. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2665 Supplier Medchemexpress LLC Supplier No. HY11109100MG
Medchemexpress, HY-11109 100mg TAK-242 Resatorvid CAS:243984-11-4 Formula:C15H17ClFNO4S TLR4 Purity:98% TAK-242 is a potent TLR4 signaling inhibitor, selectively inhibits the TLR4-mediated production of cytokines and NO. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.