Learn More
Medchemexpress LLC HY-100487 50mg , TAK-243 CAS:1450833-55-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10048750MG
Encompass_Preferred
HY-100487 50mg TAK-243 CAS: 1450833-55-2 TAK-243 (MLN7243) is a first-in-class, selective ubiquitin activating enzyme, UAE (UBA1) inhibitor (IC50=1 nM), which blocks ubiquitin conjugation, disrupting monoubiquitin signaling as well as global protein ubiquitination. TAK-243 (MLN7243) induces endoplasmic reticulum (ER) stress, abrogates NF-κB pathway activation and promotes apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9414