Learn More
Medchemexpress LLC Tak-243 (MLN7243) | 1450833-55-2 | MFCD28964160 | >98.0% | 519.5 g/mol | C19H20F3N5O5S2 | 100 MG
SDP
Catalog No. 5000213969
Encompass_Preferred
TAK-243 (MLN7243) is a potent, selective small-molecule inhibitor of the ubiquitin-activating enzyme UBA1 that blocks ubiquitin conjugation. It is used as a research reagent in studies of the ubiquitin-proteasome system and preclinical oncology models.
  • Potent, selective inhibitor of ubiquitin-activating enzyme (UBA1).
  • Low nanomolar potency (IC50 ≈ 1 nM) in enzymatic assays.
  • High chemical purity suitable for research applications.
  • Characterized molecular weight 519.5 g/mol and formula C19H20F3N5O5S2.
  • Store at -20 °C for long-term stability.
Catalog No. 50-002-13969 Supplier Medchemexpress LLC Supplier No. HY100487100MG
TAK-243 (MLN7243) is a potent, selective small-molecule inhibitor of the ubiquitin-activating enzyme UBA1 that blocks ubiquitin conjugation. It is used as a research reagent in studies of the ubiquitin-proteasome system and preclinical oncology models.
  • Potent, selective inhibitor of ubiquitin-activating enzyme (UBA1).
  • Low nanomolar potency (IC50 ≈ 1 nM) in enzymatic assays.
  • High chemical purity suitable for research applications.
  • Characterized molecular weight 519.5 g/mol and formula C19H20F3N5O5S2.
  • Store at -20 °C for long-term stability.