Learn More
Selleck Chemical LLC TAK-243 (MLN7243) 2mg 1450833-55-2
SDP
Supplier:  Selleck Chemical LLC S83412MG
Encompass_Preferred
TAK-243 (MLN7243) is a potent, mechanism-based small-molecule inhibitor of the ubiquitin activating enzyme (UAE) with an IC50 of 1 ¿ 0.2 nM in the UBCH10 E2 thioester assay. It has minimal inhibitory activity in a panel of kinase and receptor assays, as well as on human carbonic anhydrase type I and type II. TAK-243 (MLN7243) induces ER stress, abrogates NF?B pathway activation and promotes apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8193