Learn More
Medchemexpress LLC HY-100487 10mg Medchemexpress, TAK-243 MLN7243;AOB87172 CAS:1450833-55-2 Purity:98%
SDP
Catalog No. 501636756
Encompass_Preferred
Medchemexpress,HY-100487 10mg TAK-243 MLN7243;AOB87172 CAS:1450833-55-2 Formula:C19H20F3N5O5S2 TAK-243 is a potent and selective ubiquitin-like modifier activating enzyme 1 (UBA1) inhibitor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6756 Supplier Medchemexpress LLC Supplier No. HY10048710MG
Medchemexpress,HY-100487 10mg TAK-243 MLN7243;AOB87172 CAS:1450833-55-2 Formula:C19H20F3N5O5S2 TAK-243 is a potent and selective ubiquitin-like modifier activating enzyme 1 (UBA1) inhibitor. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.