Learn More
Selleck Chemical LLC TAK-285 S2784-5mg
SDP
Catalog No. 503112471
Encompass_Preferred
TAK-285 is a novel dual HER2 and EGFR(HER1) inhibitor with IC50 of 17 nM and 23 nM 10-fold selectivity for HER1/2 than HER4 less potent to MEK1/5 c-Met Aurora B Lck CSK etc Phase 1
Catalog No. 50-311-2471 Supplier Selleck Chemical LLC Supplier No. S27845MG
TAK-285 is a novel dual HER2 and EGFR(HER1) inhibitor with IC50 of 17 nM and 23 nM 10-fold selectivity for HER1/2 than HER4 less potent to MEK1/5 c-Met Aurora B Lck CSK etc Phase 1