Learn More
Medchemexpress LLC TAK-285 | 871026-44-7 | C26H25ClF3N5O3 | 50 MG
SDP
Catalog No. 5000373301
Encompass_Preferred
TAK-285 is a potent, selective, ATP-competitive, and orally active inhibitor of HER2 and EGFR (HER1), with IC50 values of 17 nM and 23 nM, respectively. It demonstrates greater than 10-fold selectivity for HER1/2 compared to HER4, and exhibits less potency against MEK1/5, c-Met, Aurora B, Lck, and CSK, among others. This compound has shown effective antitumor activity and can cross the blood-brain barrier (BBB).
  • Potent and selective inhibition
  • ATP-competitive
  • Orally active
  • Antitumor activity
  • Blood-brain barrier penetration
  • Inhibitory activity against BT-474 cells
  • Preclinical efficacy in xenograft models
Catalog No. 50-003-73301 Supplier Medchemexpress LLC Supplier No. HY1519650MG
TAK-285 is a potent, selective, ATP-competitive, and orally active inhibitor of HER2 and EGFR (HER1), with IC50 values of 17 nM and 23 nM, respectively. It demonstrates greater than 10-fold selectivity for HER1/2 compared to HER4, and exhibits less potency against MEK1/5, c-Met, Aurora B, Lck, and CSK, among others. This compound has shown effective antitumor activity and can cross the blood-brain barrier (BBB).
  • Potent and selective inhibition
  • ATP-competitive
  • Orally active
  • Antitumor activity
  • Blood-brain barrier penetration
  • Inhibitory activity against BT-474 cells
  • Preclinical efficacy in xenograft models