Learn More
Apexbio Technology LLC TAK-438 1260141-27-2 200mg
SDP
Catalog No. 5000096635
Encompass_Preferred
TAK-438 is a small-molecule inhibitor targeting gastric H K -ATPase It is designed to reversibly block potassium-competitive activity of this enzyme thereby suppressing gastric acid secretion TAK-438 exerts its biological activity primarily through direct potassium-competitive inhibition of the gastric H K -ATPase enzyme In animal studies TAK-438 demonstrates concentration-dependent inhibition of histamine-stimulated acid secretion in rats and dogs with effects persisting beyond 24 hours after oral administration Based on these pharmacological properties TAK-438 holds research potential in studies involving gastric acid physiology and acid-related gastrointestinal disorders
Catalog No. 50-000-96635 Supplier Apexbio Technology LLC Supplier No. A2308200
TAK-438 is a small-molecule inhibitor targeting gastric H K -ATPase It is designed to reversibly block potassium-competitive activity of this enzyme thereby suppressing gastric acid secretion TAK-438 exerts its biological activity primarily through direct potassium-competitive inhibition of the gastric H K -ATPase enzyme In animal studies TAK-438 demonstrates concentration-dependent inhibition of histamine-stimulated acid secretion in rats and dogs with effects persisting beyond 24 hours after oral administration Based on these pharmacological properties TAK-438 holds research potential in studies involving gastric acid physiology and acid-related gastrointestinal disorders