Learn More
Medchemexpress LLC HY-P0076A 1mg Medchemexpress, TAK-448 (acetate) CAS:1470374-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0076A1MG
Encompass_Preferred
Medchemexpress, HY-P0076A 1mg TAK-448 (acetate) CAS:1470374-22-1 TAK-448 acetate (MVT-602 acetate) is a potent and full KISS1R agonist with an IC 50 of 460 pM and an EC 50 of 632 pM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4011