Learn More
Medchemexpress LLC HY-P0076A 5mg , TAK-448 (acetate) CAS:1470374-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0076A5MG
Encompass_Preferred
HY-P0076A 5mg TAK-448 (acetate) CAS: 1470374-22-1 TAK-448 acetate (MVT-602 acetate) is a potent and full KISS1R agonist with an IC50 of 460 pM and an EC50 of 632 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9281