Learn More
TARGETMOL CHEMICALS INC TAK-580 5MG
SDP
Catalog No. 502880472
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAK-580 (MLN2480) is an oral, selective pan-Raf kinase inhibitor in Clinicalal trials. Purity 99.25%
Catalog No. 50-288-0472 Supplier TARGETMOL CHEMICALS INC Supplier No. T68955MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAK-580 (MLN2480) is an oral, selective pan-Raf kinase inhibitor in Clinicalal trials. Purity 99.25%