Learn More
TARGETMOL CHEMICALS INC TAK-593 10MG
SDP
Catalog No. 502877384
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. TAK-593 is an effective VEGFR and PDGFR family inhibitor (IC50s 3.2, 0.95, 1.1, 4.3, and 13 nM for VEGFR1, VEGFR2, VEGFR3, PDFGRalpha, and PDFGRbeta, respectively). Purity 99.18%
Catalog No. 50-287-7384 Supplier TARGETMOL CHEMICALS INC Supplier No. T1697510MG
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. TAK-593 is an effective VEGFR and PDGFR family inhibitor (IC50s 3.2, 0.95, 1.1, 4.3, and 13 nM for VEGFR1, VEGFR2, VEGFR3, PDFGRalpha, and PDFGRbeta, respectively). Purity 99.18%