Learn More
Selleck Chemical LLC TAK-632
SDP
Catalog No. 501364602
Encompass_Preferred
TAK-632 is a potent pan-Raf inhibitor with IC50 of 8 3 nM and 1 4 nM for B-Raf(wt) and C-Raf in cell-free assays respectively showing less or no inhibition against other tested kinases
Catalog No. 50-136-4602 Supplier Selleck Chemical LLC Supplier No. S72915MG
TAK-632 is a potent pan-Raf inhibitor with IC50 of 8 3 nM and 1 4 nM for B-Raf(wt) and C-Raf in cell-free assays respectively showing less or no inhibition against other tested kinases