Learn More
Selleck Chemical LLC TAK-659 2mg 1952251-28-3
SDP
Supplier:  Selleck Chemical LLC S84422MG
Encompass_Preferred
TAK-659 is a potent and selective inhibitor of spleen tyrosine kinase (SYK) with an IC50 value of 3.2 nM. It is selective against most other kinases, but potent toward both SYK and FLT3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8137