Learn More
Medchemexpress LLC HY-100867A 100mg , TAK-659 (hydrochloride) CAS:1952251-28-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY100867A100MG
Encompass_Preferred
Medchemexpress, HY-100867A 100mg TAK-659 (hydrochloride) CAS:1952251-28-3 Formula:C17H22ClFN6O IC50: 3.2 nM (Syk) Purity:98% TAK-659 hydrochloride is a potent, selective and orally available spleen tyrosine kinase ( Syk ) inhibitor with an IC 50 of 3.2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3247