Learn More
Selleck Chemical LLC TAK-659 Hydrochloride
SDP
Catalog No. 5000001863
Encompass_Preferred
TAK-659 Hydrochloride is a potent and selective inhibitor of spleen tyrosine kinase (SYK) with an IC50 value of 3 2 nM It is selective against most other kinases but potent toward both SYK and FLT3
Catalog No. 50-000-01863 Supplier Selleck Chemical LLC Supplier No. S84425MG
TAK-659 Hydrochloride is a potent and selective inhibitor of spleen tyrosine kinase (SYK) with an IC50 value of 3 2 nM It is selective against most other kinases but potent toward both SYK and FLT3