Learn More
Medchemexpress LLC Tak-683 acetate | 99.9% | 1358.50 | 10 MG
SDP
Catalog No. 5000424149
Encompass_Preferred
TAK-683 acetate is a potent full KISS1 receptor (KISS1R) agonist. It is a nonapeptide metastin analog with improved metabolic stability, showing agonistic activities to KISS1R. It depletes GnRH in the hypothalamus and reduces plasma FSH, LH, and testosterone levels in vivo, suggesting potential for the study of hormone-dependent prostate cancer.
  • Potent full KISS1 receptor agonist
  • Improved metabolic stability
  • Nonapeptide metastin analog
  • Depletes GnRH in the hypothalamus
  • Reduces plasma FSH, LH, and testosterone levels in vivo
  • Potential for the study of hormone-dependent prostate cancer
  • Exhibits IC50 value of 150-180 pM in rat KISS1R-expressing CHO cells
Catalog No. 50-004-24149 Supplier Medchemexpress LLC Supplier No. HYP2161B10MG
TAK-683 acetate is a potent full KISS1 receptor (KISS1R) agonist. It is a nonapeptide metastin analog with improved metabolic stability, showing agonistic activities to KISS1R. It depletes GnRH in the hypothalamus and reduces plasma FSH, LH, and testosterone levels in vivo, suggesting potential for the study of hormone-dependent prostate cancer.
  • Potent full KISS1 receptor agonist
  • Improved metabolic stability
  • Nonapeptide metastin analog
  • Depletes GnRH in the hypothalamus
  • Reduces plasma FSH, LH, and testosterone levels in vivo
  • Potential for the study of hormone-dependent prostate cancer
  • Exhibits IC50 value of 150-180 pM in rat KISS1R-expressing CHO cells