Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAK-683 acetate | 99.9% | 1358.50 | 5 MG
SDP

Catalog No. 5000422990
Encompass_Preferred

TAK-683 acetate is a potent full KISS1 receptor (KISS1R) agonist with improved metabolic stability. This nonapeptide metastin analog exhibits agonistic activities to KISS1R and can deplete GnRH in the hypothalamus, reducing plasma FSH, LH, and testosterone levels in vivo. It has potential for studying hormone-dependent prostate cancer.

  • Potent full KISS1 receptor (KISS1R) agonist (IC50=170 pM)
  • Improved metabolic stability
  • Nonapeptide metastin analog
  • Exhibits agonistic activities to KISS1R with EC50 values of 0.96 nM for human and 1.6 nM for rat
  • Depletes GnRH in the hypothalamus
  • Reduces plasma FSH, LH, and testosterone levels in vivo
  • Potential for the study of hormone-dependent prostate cancer
  • Molecular weight: 1358.50
  • Target: Kisspeptin receptor
  • Pathway: GPCR/G protein

Catalog No. 50-004-22990 Supplier Medchemexpress LLC Supplier No. HYP2161B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TAK-683 acetate is a potent full KISS1 receptor (KISS1R) agonist with improved metabolic stability. This nonapeptide metastin analog exhibits agonistic activities to KISS1R and can deplete GnRH in the hypothalamus, reducing plasma FSH, LH, and testosterone levels in vivo. It has potential for studying hormone-dependent prostate cancer.

  • Potent full KISS1 receptor (KISS1R) agonist (IC50=170 pM)
  • Improved metabolic stability
  • Nonapeptide metastin analog
  • Exhibits agonistic activities to KISS1R with EC50 values of 0.96 nM for human and 1.6 nM for rat
  • Depletes GnRH in the hypothalamus
  • Reduces plasma FSH, LH, and testosterone levels in vivo
  • Potential for the study of hormone-dependent prostate cancer
  • Molecular weight: 1358.50
  • Target: Kisspeptin receptor
  • Pathway: GPCR/G protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.