Learn More
Medchemexpress LLC HY-13449 5mg Medchemexpress, TAK-733 CAS:1035555-63-5 Purity:>98%
SDP
Catalog No. 502030313
Encompass_Preferred
Medchemexpress, HY-13449 5mg TAK-733 CAS:1035555-63-5 TAK-733 is a potent and selective MEK allosteric site inhibitor with an IC50 of 3.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0313 Supplier Medchemexpress LLC Supplier No. HY134495MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-13449 5mg TAK-733 CAS:1035555-63-5 TAK-733 is a potent and selective MEK allosteric site inhibitor with an IC50 of 3.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.