Learn More
TARGETMOL CHEMICALS INC TAK-779 10MG
SDP
Catalog No. 502876564
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAK-779 (Takeda 779) is an antagonist of chemokine receptor 5 (CCR5), CCR2b, and CXC chemokine receptor 3 (CXCR3). Purity 99.58%
Catalog No. 50-287-6564 Supplier TARGETMOL CHEMICALS INC Supplier No. T749910MG
Add to Cart
Add to Cart
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAK-779 (Takeda 779) is an antagonist of chemokine receptor 5 (CCR5), CCR2b, and CXC chemokine receptor 3 (CXCR3). Purity 99.58%