Learn More
Apexbio Technology LLC TAK-875, 10mg. Cas: 1000413-72-8 MFCD: MFCD18251445. GPR40 agonist
SDP
Supplier:  Apexbio Technology LLC A833910
Encompass_Preferred
TAK-875 is a potent, selective, and oral GPR40 agonist. GPR40 is one of the G protein-coupled receptors predominantly expressed in pancreatic -cells, mediating enhancement of glucose-stimulated insulin secretion by free fatty acids. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5116