Learn More
Apexbio Technology LLC TAK-901 934541-31-8 200mg
SDP
Catalog No. 5000095349
Encompass_Preferred
TAK-901 (CAS 934541-31-8) is a small-molecule inhibitor primarily targeting Aurora B kinase with reported IC50 values of 0 0017 M for Aurora-B/INCENP and 0 021 M for Aurora-A/TPX2 Derived from an azacarboline kinase hinge-binding chemotype TAK-901 demonstrates time-dependent selective inhibition of Aurora B suppressing histone H3 phosphorylation and inducing polyploidy in vitro It inhibits proliferation across a range of human cancer cell lines (40 500 nM) and additionally targets kinases such as FLT3 and FGFR2 in cells In preclinical rodent xenograft models TAK-901 reduces tumor growth and induces complete regression in ovarian cancer models A phase I clinical trial has evaluated its dosing and pharmacokinetic characteristics in patients with advanced solid tumors and lymphomas
Catalog No. 50-000-95349 Supplier Apexbio Technology LLC Supplier No. A4124200
TAK-901 (CAS 934541-31-8) is a small-molecule inhibitor primarily targeting Aurora B kinase with reported IC50 values of 0 0017 M for Aurora-B/INCENP and 0 021 M for Aurora-A/TPX2 Derived from an azacarboline kinase hinge-binding chemotype TAK-901 demonstrates time-dependent selective inhibition of Aurora B suppressing histone H3 phosphorylation and inducing polyploidy in vitro It inhibits proliferation across a range of human cancer cell lines (40 500 nM) and additionally targets kinases such as FLT3 and FGFR2 in cells In preclinical rodent xenograft models TAK-901 reduces tumor growth and induces complete regression in ovarian cancer models A phase I clinical trial has evaluated its dosing and pharmacokinetic characteristics in patients with advanced solid tumors and lymphomas