Learn More
Medchemexpress LLC TAK-960 dihydrochloride | 99.2% | 634.52 | 5 MG
SDP
Catalog No. 5000364364
Encompass_Preferred
TAK-960 dihydrochloride is an orally available, selective inhibitor of polo-like kinase 1 (PLK1), with an IC50 of 0.8 nM. It also demonstrates inhibitory activities against PLK2 and PLK3, with IC50s of 16.9 and 50.2 nM, respectively. This compound inhibits the proliferation of multiple cancer cell lines and shows significant efficacy against various tumor xenografts.
  • Orally available
  • Selective inhibitor of polo-like kinase 1 (PLK1)
  • Inhibits proliferation of multiple cancer cell lines
  • Exhibits significant efficacy against various tumor xenografts
  • Causes accumulation of G2-M cells
  • Leads to G2/M cell cycle arrest
Catalog No. 50-003-64364 Supplier Medchemexpress LLC Supplier No. 5MGHY15160B
TAK-960 dihydrochloride is an orally available, selective inhibitor of polo-like kinase 1 (PLK1), with an IC50 of 0.8 nM. It also demonstrates inhibitory activities against PLK2 and PLK3, with IC50s of 16.9 and 50.2 nM, respectively. This compound inhibits the proliferation of multiple cancer cell lines and shows significant efficacy against various tumor xenografts.
  • Orally available
  • Selective inhibitor of polo-like kinase 1 (PLK1)
  • Inhibits proliferation of multiple cancer cell lines
  • Exhibits significant efficacy against various tumor xenografts
  • Causes accumulation of G2-M cells
  • Leads to G2/M cell cycle arrest