Learn More
Medchemexpress LLC HY-111789 5mg Medchemexpress, TAK-981 CAS:1858276-04-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1117895MG
Encompass_Preferred
Medchemexpress, HY-111789 5mg TAK-981 CAS:1858276-04-6 TAK-981 is a first in class and selective inhibitor of the SUMOylation enzymatic cascade, with potential immune-activating and antineoplastic activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9701