Learn More
Medchemexpress LLC Tak1 protein, human (Sf9) | ≥98.0% | 20UG
SDP
Catalog No. 5000174631
Encompass_Preferred
Recombinant human TAK1 protein produced in Sf9 insect cells, supplied as a tag-free, highly purified preparation suitable for biochemical assays, kinase activity studies, and antibody validation. Provided in small-scale quantities; aliquot and store frozen to preserve activity.
  • Tag-free recombinant human TAK1 protein.
  • Expressed in Sf9 insect cells for eukaryotic post-translational modifications.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level for sensitive applications.
  • Supplied at a concentration that facilitates small-volume assays.
  • Stable when stored at -80°C; avoid repeated freeze-thaw cycles.
Catalog No. 50-001-74631 Supplier Medchemexpress LLC Supplier No. HYP70359120UG
Recombinant human TAK1 protein produced in Sf9 insect cells, supplied as a tag-free, highly purified preparation suitable for biochemical assays, kinase activity studies, and antibody validation. Provided in small-scale quantities; aliquot and store frozen to preserve activity.
  • Tag-free recombinant human TAK1 protein.
  • Expressed in Sf9 insect cells for eukaryotic post-translational modifications.
  • High purity suitable for biochemical and cell-based assays.
  • Low endotoxin level for sensitive applications.
  • Supplied at a concentration that facilitates small-volume assays.
  • Stable when stored at -80°C; avoid repeated freeze-thaw cycles.