Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAK1/MAP4K2 inhibitor 1 | 1315330-11-0 | 99.9% | C29H31F3N6O2 | 1 ML
SDP

Catalog No. 5000435082
Encompass_Preferred

TAK1/MAP4K2 inhibitor 1 is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. It is identified by CAS number 1315330-11-0 and has a chemical formula of C29H31F3N6O2. This inhibitor appears as a white to light yellow solid with a purity of 99.9%.

  • Used for laboratory research applications.
  • Solid appearance, white to light yellow.
  • High purity of 99.9%.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping at room temperature for less than 2 weeks.

Catalog No. 50-004-35082 Supplier Medchemexpress LLC Supplier No. HY7725110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TAK1/MAP4K2 inhibitor 1 is a chemical compound primarily used as a laboratory chemical and for the manufacture of substances. It is identified by CAS number 1315330-11-0 and has a chemical formula of C29H31F3N6O2. This inhibitor appears as a white to light yellow solid with a purity of 99.9%.

  • Used for laboratory research applications.
  • Solid appearance, white to light yellow.
  • High purity of 99.9%.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping at room temperature for less than 2 weeks.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.