Learn More
Medchemexpress LLC Tak1/map4k2 inhibitor 1 | 1315330-11-0 | 99.9% | C29H31F3N6O2 | 50 MG
SDP
Catalog No. 5000434975
Encompass_Preferred
TAK1/MAP4K2 inhibitor 1 is a potent dual TGFβ-activated kinase 1 (TAK1) and mitogen-activated protein kinase kinase kinase kinase 2 (MAP4K2) inhibitor. It exhibits IC50s of 41.1 nM for TAK1 and 18.2 nM for MAP4K2.
  • Potent dual inhibitor of TAK1 and MAP4K2
  • IC50 of 41.1 nM for TAK1
  • IC50 of 18.2 nM for MAP4K2
  • Exhibits antiproliferative activity against parental mouse BaF3 cells harbouring Bcr/Abl
  • Moderate terminal elimination half-life in mice
  • Good solubility in DMSO
Catalog No. 50-004-34975 Supplier Medchemexpress LLC Supplier No. HY7725150MG
TAK1/MAP4K2 inhibitor 1 is a potent dual TGFβ-activated kinase 1 (TAK1) and mitogen-activated protein kinase kinase kinase kinase 2 (MAP4K2) inhibitor. It exhibits IC50s of 41.1 nM for TAK1 and 18.2 nM for MAP4K2.
  • Potent dual inhibitor of TAK1 and MAP4K2
  • IC50 of 41.1 nM for TAK1
  • IC50 of 18.2 nM for MAP4K2
  • Exhibits antiproliferative activity against parental mouse BaF3 cells harbouring Bcr/Abl
  • Moderate terminal elimination half-life in mice
  • Good solubility in DMSO