Learn More
Enzo Life Sciences TAK1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKASST009E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse TAK1. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7321