Learn More
Eurofins Discoverx TAK1-TAB1 FUSION, active, 10 µg. (14-600)
Supplier:  Eurofins Discoverx 14600
Encompass
N-terminal 6xHis-tagged recombinant, human TAK1 amino acids 1-303, fused to human TAB1 amino acids 437-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9029