Learn More
Cayman Chemical TAK1-TAB1 FusIonhumnreco 10ug
SDP
Catalog No. 502950403
Encompass_Preferred
Source: Active recombinant N-terminal GST-tagged human TAK1 fused to TAB1 via a DFGGGGG peptide linker expressed in insect cells • Amino acids: TAK1 (1-303) and TAB1 (437-504) • MW: 69 kDa
Catalog No. 50-295-0403 Supplier Cayman Chemical Supplier No. 3399010 UG
Source: Active recombinant N-terminal GST-tagged human TAK1 fused to TAB1 via a DFGGGGG peptide linker expressed in insect cells • Amino acids: TAK1 (1-303) and TAB1 (437-504) • MW: 69 kDa