Learn More
Takara Bio TaKaRa Ex Taq® DNA Polymerase, 250 units
SDP
Catalog No. 50443968
Encompass
TaKaRa Ex Taq DNA Polymerase combines the proven performance of Takara Taq polymerase with the proofreading activity of an efficient 3'-to-5' exonuclease, for high-sensitivity, high-efficiency PCR. Ex Taq can also be used for long-range PCR (up to 20 kb from genomic DNA templates and up to 30 kb from lambda DNA templates). Separate tubes of Mg2+ plus buffer and dNTP mix are supplied with the polymerase. 250 units
Catalog No. 50-443-968 Supplier Takara Bio Supplier No. RR001A
TaKaRa Ex Taq DNA Polymerase combines the proven performance of Takara Taq polymerase with the proofreading activity of an efficient 3'-to-5' exonuclease, for high-sensitivity, high-efficiency PCR. Ex Taq can also be used for long-range PCR (up to 20 kb from genomic DNA templates and up to 30 kb from lambda DNA templates). Separate tubes of Mg2+ plus buffer and dNTP mix are supplied with the polymerase. 250 units