Learn More
Anton Paar Take DMA 35 anywhere and measure density and concentration directly on-site. This portable meter replaces glass hydrometers, uses only 2 mL of sample, and delivers lab-level accuracy. Built for field use, it includes RFID and Bluetooth for easy data handling.