Learn More
Medchemexpress LLC HY-103490 10mg Medchemexpress, Takinib CAS:1111556-37-6 Purity:>98%
SDP
Catalog No. 502028730
Encompass_Preferred
Medchemexpress, HY-103490 10mg Takinib CAS:1111556-37-6 Takinib is a potent and selective TAK1 inhibitor with an IC50 of 9.5 nM, which inhibits autophosphorylated and non-phosphorylated TAK1 that binds within the ATP-binding pocket and inhibits by slowing down the rate-limiting step of TAK1 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8730 Supplier Medchemexpress LLC Supplier No. HY10349010MG
Medchemexpress, HY-103490 10mg Takinib CAS:1111556-37-6 Takinib is a potent and selective TAK1 inhibitor with an IC50 of 9.5 nM, which inhibits autophosphorylated and non-phosphorylated TAK1 that binds within the ATP-binding pocket and inhibits by slowing down the rate-limiting step of TAK1 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.