Learn More
Selleck Chemical LLC Takinib 5mg 1111556-37-6 EDHS-206
SDP
Supplier:  Selleck Chemical LLC S86635MG
Encompass_Preferred
Takinib (EDHS-206) is a potent and selective TAK1 inhibitor with an IC50 of 9.5 nM, more than 1.5 log more potent than the second and third ranked targets, IRAK4 (120 nM) and IRAK1 (390 nM), respectively. Takinib induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8048