Learn More
Aviva Systems Biology Corp TAL1 Antibody - middle region (P100970_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TAL1. Alternate names: SCL, TCL5, tal-1, bHLHa17ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53810