Learn More
Medchemexpress LLC HY-13233B 5mg Medchemexpress, Talabostat isomer mesylate CAS: Purity:>98%
SDP
Catalog No. 501932655
Encompass_Preferred
Medchemexpress, HY-13233B 5mg Talabostat isomer mesylate CAS: Talabostat isomer mesylate is an isomer of talabostat mesylate. Talabostat (PT100, Val-boroPro) is a potent, nonselective and orally available dipeptidyl peptidase IV (DPP-IV) inhibitor with a K i of 0.18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2655 Supplier Medchemexpress LLC Supplier No. HY13233B5MG
Medchemexpress, HY-13233B 5mg Talabostat isomer mesylate CAS: Talabostat isomer mesylate is an isomer of talabostat mesylate. Talabostat (PT100, Val-boroPro) is a potent, nonselective and orally available dipeptidyl peptidase IV (DPP-IV) inhibitor with a K i of 0.18 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.