Learn More
Medchemexpress LLC HY-13233A 50mg , Talabostat (mesylate) Val-boroPro;PT100 CAS:150080-09-4 Purity:98%
SDP
Catalog No. 501493623
Encompass_Preferred
Medchemexpress, HY-13233A 50mg Talabostat (mesylate) Val-boroPro;PT100 CAS:150080-09-4 Formula:C10H23BN2O6S Ki: 0.18 nM (DPP-IV), 1.5 nM (DPP8), 0.76 nM (DPP9) Purity:98% Talabostat mesylate is a potent, nonselective and orally available dipeptidyl peptidase IV (DPP-IV) inhibitor with a K i of 0.18 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3623 Supplier Medchemexpress LLC Supplier No. HY13233A50MG
Medchemexpress, HY-13233A 50mg Talabostat (mesylate) Val-boroPro;PT100 CAS:150080-09-4 Formula:C10H23BN2O6S Ki: 0.18 nM (DPP-IV), 1.5 nM (DPP8), 0.76 nM (DPP9) Purity:98% Talabostat mesylate is a potent, nonselective and orally available dipeptidyl peptidase IV (DPP-IV) inhibitor with a K i of 0.18 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.