Learn More
Apexbio Technology LLC Talabostat mesylate 150080-09-4 10mg
SDP
Catalog No. 5000063077
Encompass_Preferred
Talabostat mesylate (CAS 150080-09-4) also known as PT-100 or Val-boroPro is an orally active inhibitor targeting dipeptidyl peptidase enzymes particularly dipeptidyl peptidase IV (DPP4) and fibroblast activation protein (FAP) It exhibits potent inhibition against these post-prolyl proteases with an IC50 of approximately 1 nM By blocking the enzymatic cleavage of N-terminal Xaa-Pro or Xaa-Ala sequences talabostat mesylate stimulates cytokine and chemokine production enhancing hematopoiesis through induction of colony-stimulating factors such as granulocyte colony-stimulating factor (G-CSF) Due to these activities talabostat serves as a valuable pharmacological tool in studying tumor biology immune modulation and enzyme-mediated signaling pathways
Catalog No. 50-000-63077 Supplier Apexbio Technology LLC Supplier No. B394110
Talabostat mesylate (CAS 150080-09-4) also known as PT-100 or Val-boroPro is an orally active inhibitor targeting dipeptidyl peptidase enzymes particularly dipeptidyl peptidase IV (DPP4) and fibroblast activation protein (FAP) It exhibits potent inhibition against these post-prolyl proteases with an IC50 of approximately 1 nM By blocking the enzymatic cleavage of N-terminal Xaa-Pro or Xaa-Ala sequences talabostat mesylate stimulates cytokine and chemokine production enhancing hematopoiesis through induction of colony-stimulating factors such as granulocyte colony-stimulating factor (G-CSF) Due to these activities talabostat serves as a valuable pharmacological tool in studying tumor biology immune modulation and enzyme-mediated signaling pathways