Learn More
Cayman Chemical Talabostatmesylate 50mg
SDP
Catalog No. 502938433
Encompass_Preferred
A non-selective inhibitor of DPPs (IC50s = >4, 310, 4, 11, 560, and 390 nM for DPP-4, DPP-7, DPP-8, DPP-9, FAP, and PREP, respectively); inhibits proliferation of superantigen-stimulated human PBMCs (IC50 = ~10 nM); increases expression of a variety of cytokines and chemokines in tumors and tumor-draining lymph nodes in a WEHI-164 fibrosarcoma mouse model at 5 μg twice per day; reduces tumor growth in a variety of syngeneic mouse models; increases the efficacy of the antitumor antibodies rituximab and trastuzumab in Namalwa B cell lymphoma and LS180 colon carcinoma mouse xenograft models, respectively
Catalog No. 50-293-8433 Supplier Cayman Chemical Supplier No. 2900750 MG
A non-selective inhibitor of DPPs (IC50s = >4, 310, 4, 11, 560, and 390 nM for DPP-4, DPP-7, DPP-8, DPP-9, FAP, and PREP, respectively); inhibits proliferation of superantigen-stimulated human PBMCs (IC50 = ~10 nM); increases expression of a variety of cytokines and chemokines in tumors and tumor-draining lymph nodes in a WEHI-164 fibrosarcoma mouse model at 5 μg twice per day; reduces tumor growth in a variety of syngeneic mouse models; increases the efficacy of the antitumor antibodies rituximab and trastuzumab in Namalwa B cell lymphoma and LS180 colon carcinoma mouse xenograft models, respectively