Learn More
Selleck Chemical LLC Taladegib (LY2940680) S2157-5mg
SDP
Catalog No. 503111345
Encompass_Preferred
Taladegib (LY2940680) binds to the Smoothened (Smo) receptor and potently inhibits Hedgehog (Hh) signaling Phase 1/2
Catalog No. 50-311-1345 Supplier Selleck Chemical LLC Supplier No. S21575MG
Taladegib (LY2940680) binds to the Smoothened (Smo) receptor and potently inhibits Hedgehog (Hh) signaling Phase 1/2