Learn More
Medchemexpress LLC HY-13242 5mg Medchemexpress, Taladegib CAS:1258861-20-9 Purity:>98%
SDP
Catalog No. 502081312
Encompass_Preferred
Medchemexpress, HY-13242 5mg Taladegib CAS:1258861-20-9 Taladegib (LY2940680) is an antagonist of the smoothened receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1312 Supplier Medchemexpress LLC Supplier No. HY132425MG
Medchemexpress, HY-13242 5mg Taladegib CAS:1258861-20-9 Taladegib (LY2940680) is an antagonist of the smoothened receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.