Learn More
Medchemexpress LLC Talaporfin sodium | 220201-34-3 | 99.7% | 799.69 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1647710MM/1ML
Encompass_Preferred
Talaporfin sodium, also known as ME2906, is a chlorin-based photosensitizer used for research into various cancers through photodynamic therapy (PDT).
  • Appearance: Solid
  • Color: Purple to black
  • For research use only
Catalog No. 50-003-94082