Learn More
Cayman Chemical Talaporfin (sodium salt), Purity: ≥95%, CAS: 220201-34-3, 10 mg
SDP
Catalog No. 502911398
Encompass_Preferred
A photosensitizer; induces apoptosis, DNA DSBs, and the production of ROS in TE-11R 5-fluorouracil-resistant cells at 30 µg/ml in combination with radiation; reduces tumor growth in a TE-11R ESCC mouse xenograft model at 2.5, 5, and 10 mg/kg in combination with PDT
Catalog No. 50-291-1398 Supplier Cayman Chemical Supplier No. 3572210 MG
A photosensitizer; induces apoptosis, DNA DSBs, and the production of ROS in TE-11R 5-fluorouracil-resistant cells at 30 µg/ml in combination with radiation; reduces tumor growth in a TE-11R ESCC mouse xenograft model at 2.5, 5, and 10 mg/kg in combination with PDT