Learn More
ATCC Levilactobacillus brevis; Strain 118-8; genomic DNA
Catalog No. 502381302
Encompass
Genomic DNA isolated from Levilactobacillus brevis strain 118-8 (ATCC 367). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-1302 Supplier ATCC Supplier No. 367D5
Genomic DNA isolated from Levilactobacillus brevis strain 118-8 (ATCC 367). This whole-genome sequenced product can be used in PCR and other molecular biology applications.